missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat anti-Mouse IgG (Heavy chain) Secondary Antibody, Biotin
Description
SA1-25198 detects mouse IgG1, Heavy Chain.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spécifications
Spécifications
| Antigène | Mouse IgG (Heavy chain) |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | LO-MG1-2 |
| Concentration | 1 mg/mL |
| Conjugué | Biotin |
| Formule | PBS with 50% glycerol and 0.1% sodium azide |
| Espèces hôtes | Rat |
| Immunogène | Purified mouse IgG1 |
| Méthode de purification | Antigen affinity chromatography |
| Afficher plus de résultats |