missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat anti-Mouse IgE (Heavy chain) Secondary Antibody, Biotin

Catalog No. PIMA516779
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516779 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516779 Supplier Invitrogen™ Supplier No. MA516779
  • ONLINE EXCLUSIVE PRICE
    $1,060.43
    $616.50 / Each

    Save $443.93

    42.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Secondary Antibody

This antibody does not react with rat. Rat anti Mouse IgE Heavy Chain antibody, clone LO-ME-3 recognizes the murine epsilon immunoglobulin heavy chain, and does not cross react with other murine immunoglobulin classes or subclasses.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Mouse IgE (Heavy chain)
Applications ELISA
Classification Monoclonal
Clone LO-ME-3
Concentration 1 mg/mL
Conjugate Biotin
Formulation PBS with 50% glycerol and 0.1% sodium azide
Host Species Rat
Immunogen Purified mouse IgE.
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less