missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RASSF2 Monoclonal Antibody (K94034_20H8), Invitrogen™

Catalog No. MA555700
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555700 50 μL
MA556080 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555700 Supplier Invitrogen™ Supplier No. MA555700
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: QDDNERIRPP PSSSSWHSGC NLGAQGTTLK PLTVPKVQIS EVDAPPEGDQ MPSSTDSRGL KPLQEDTPQL MRTRSDVGVR RRGNVRTPSD

Potential tumor suppressor. Acts as a KRAS-specific effector protein. May promote apoptosis and cell cycle arrest. Stabilizes STK3/MST2 by protecting it from proteasomal degradation.

Specifications

Antigen RASSF2
Applications Western Blot
Classification Monoclonal
Clone K94034_20H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene RASSF2
Gene Accession No. P50749
Gene Alias 3830431H01Rik; 9030412M04Rik; AI852669; AW495050; CENP-34; centromere protein 34; Kiaa0168; Ras association (RalGDS/AF-6) domain family 2; Ras association (RalGDS/AF-6) domain family member 2; Ras association domain family member 2; ras association domain-containing protein 2; RASFADIN; Rassf2; RP4-599I11.1
Gene Symbols RASSF2
Host Species Mouse
Immunogen Recombinant human RASSF2 (immunogen range is 71-160).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9770
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less