missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RASEF Recombinant Rabbit Monoclonal Antibody (23GB2745)

Catalog No. MA552521
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552521 20 μL
MA552520 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552521 Supplier Invitrogen™ Supplier No. MA552521

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

RASEF Recombinant Rabbit Monoclonal Antibody (23GB2745)

RASEF Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Specifications

Antigen RASEF
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2745
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RASEF
Gene Accession No. Q5RI75, Q8IZ41
Gene Alias BC030882; RAB45; RAS and EF hand domain containing; RAS and EF-hand domain containing; ras and EF-hand domain-containing protein; Ras and EF-hand domain-containing protein homolog; RASEF; ras-related protein Rab-45
Gene Symbols RASEF
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB45 (82-120AA).
Purification Method Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158158, 242505, 298138
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less