missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASEF Recombinant Rabbit Monoclonal Antibody (23GB2745)

Description
RASEF Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Spécifications
Spécifications
| Antigène | RASEF |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB2745 |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gène | RASEF |
| Numéro d’acquisition du gène | Q5RI75, Q8IZ41 |
| Alias du gène | BC030882; RAB45; RAS and EF hand domain containing; RAS and EF-hand domain containing; ras and EF-hand domain-containing protein; Ras and EF-hand domain-containing protein homolog; RASEF; ras-related protein Rab-45 |
| Symboles de gène | RASEF |
| Afficher plus de résultats |