missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RARA/RARB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115840
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115840 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115840 Supplier Invitrogen™ Supplier No. PA5115840
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Retinoic Acid Receptor alpha/beta.

Retinoic acid, a metabolite of vitamin A, is necessary for normal organogenesis but acts as a teratogen at high levels during embryonic and fetal development. Retinoic acid functions through its interaction with the nuclear protein, retinoic acid receptor (RAR). RAR belongs to the steroid and thyroid hormone superfamily of nuclear receptor proteins which exert their effects by binding to specific DNA response elements, thus regulating gene expression in target cells. RAR exists as three major subtypes: alpha, beta and gamma.

Specifications

Antigen RARA/RARB
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene RARA
Gene Accession No. P10276, P10826, P11416, P22605
Gene Alias A830025K23; HAP; HBV-activated protein; hepatitis B virus activated protein; MCOPS12; Nr1b1; Nr1b2; Nuclear receptor subfamily 1 group B member 1; nuclear receptor subfamily 1 group B member 2; nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form; RAR; RAR alpha 1; RARA; RAR-alpha; RARalpha1; Rarb; RAR-beta; RARbeta1; RAR-epsilon; retinoic acid; retinoic acid nuclear receptor alpha variant 1; retinoic acid nuclear receptor alpha variant 2; retinoic acid receptor; retinoic acid receptor alpha; Retinoic acid receptor beta; retinoic acid receptor, alpha; retinoic acid receptor, alpha polypeptide; retinoic acid receptor, beta; retinoic acid receptor, beta polypeptide; RRB2
Gene Symbols RARA, RARB
Host Species Rabbit
Immunogen A synthesized peptide derived from human RARA(Accession P10276), corresponding to amino acid residues P73-T123.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19401, 218772, 24705, 24706, 5914, 5915
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less