Learn More
Applied Biosystems™ RapidLINK™ Software v2.0
Catalog No. A59183
Change view
Click to view available options
Quantity:
Ea.
Catalog No. Quantity
A59183 Ea.
1 options
Catalog No. A59183 Supplier Applied Biosystems™ Supplier No. A59183
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Applied Biosystems RapidLINK Software v2.0 is part of the Applied Biosystems RapidHIT ID System.
The Applied Biosystems RapidLINK Software v2.0 is part of the Applied Biosystems RapidHIT ID System. This fast and simple DNA system produces lab-quality forensic DNA profiles that are aggregated within the software for direct upload to the database of choice or for further review and analysis. This data management software expedites identity confirmation securely and efficiently.
Features of the RapidLINK Software v2.0 include:
  • Securely manage one or more Applied Biosystems RapidHIT ID systems from a central location>
  • Review DNA profile results in real time prior to upload to a database
  • Enable or disable users in real time
  • Collect and review metadata using built-in reporting tools
  • Expert system-capable
  • Sample/data accessioning workflow-capable
Delivery method: After purchase you will receive a letter with upgrade/installation details.
Specifications
Product Line RapidLINK
Product Type Software
Quantity Ea.
Shipping Condition Room Temperature
For Research Use Only.