missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ RapidINTEL™ Plus Positive Control Cartridge
Description
The Applied Biosystems RapidINTEL Plus Positive Control Cartridge is for use with the Applied Biosystems RapidHIT ID System (Cat. No. A41810) to verify instrument and/or reagent performance. Each cartridge is individually wrapped and single use/disposable.
Required for use
- Applied Biosystems RapidHIT ID System (Cat. No. A41810)
- RapidHIT ID Instrument Software v2.0 or higher
- RapidHIT ID Primary Cartridge (Cat. No. A53084)
Specifications
Specifications
| Format | Cartridge |
| For Use With (Equipment) | RapidHIT™ ID |
| For Use With (Application) | Human Identification |
| Quantity | 1 Cartridge |
For Research Use Only.