missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ RapidINTEL™ Plus Ladder Cartridge Kit

Catalog No. A54344
Change view
Click to view available options
Quantity:
1 Kit (5 Cartridges)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A54344 1 Kit (5 Cartridges)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A54344 Supplier Applied Biosystems™ Supplier No. A54344

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems RapidINTEL Plus Ladder Cartridge Kit is for use with the Applied Biosystems RapidHIT ID System (Cat. No. A41810).

The Applied Biosystems RapidINTEL Plus Ladder Cartridge Kit is for use with the Applied Biosystems RapidHIT ID System (Cat. No. A41810). This kit contains five RapidINTEL Plus Ladder Cartridges for use with RapidINTEL Plus Sample Cartridges.

Required for use

  • Applied Biosystems RapidHIT ID System (Cat. No. A41810)
  • RapidHIT ID Instrument Software v2.0 or higher
  • RapidHIT ID Primary Cartridge (Cat. No. A53084)

Specifications

Format Kit
For Use With (Equipment) RapidHIT™ ID
For Use With (Application) Human Identification
Quantity 1 Kit (5 Cartridges)

For Research Use Only.