missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ RapidINTEL™ Plus 50 Sample Cartridge Kit

Catalog No. A54338
Change view
Click to view available options
Quantity:
1 Kit (50 Cartridges)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A54338 1 Kit (50 Cartridges)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A54338 Supplier Applied Biosystems™ Supplier No. A54338

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems RapidINTEL Plus 50 Sample Cartridge Kit is used with the Applied Biosystems RapidHIT ID System (Cat. No. A41810) to generate lab-quality forensic DNA profiles.

The Applied Biosystems RapidINTEL Plus 50 Sample Cartridge Kit is used with the Applied Biosystems RapidHIT ID System (Cat. No. A41810) to generate lab-quality forensic DNA profiles. The DNA profiles generated from RapidINTEL Plus sample cartridges can be compared to DNA profiles obtained from crime scene evidence to link one crime to another or to DNA profiles from reference samples to link a crime to an individual.

This kit contains 50 RapidINTEL Plus Sample Cartridges, 2 RapidINTEL Plus Positive Control Cartridges, and 2 RapidINTEL Plus Negative Control Cartridges, each of which is an individually-wrapped, single-use, disposable cartridge that contains reagents for developing a forensic DNA profile from a single sample in 90 minutes.

The RapidINTEL 'Plus' sample cartridge is an upgraded cartridge that provides more complete, more consistent data from crime scene samples. In addition, internal quality control (IQC) markers and quantification markers have been added to assist users with identification of low quantity, degraded, and inhibited samples.

To run reference samples on the Applied Biosystems RapidHIT ID System, see our ACE GlobalFiler Sample Cartridge Kit (Cat. No. A41831 or 400114RH).

Required for use

  • Applied Biosystems RapidHIT ID System (Cat. No. A41810)
  • RapidHIT ID Instrument Software v2.0 or higher
  • RapidHIT ID Primary Cartridge (Cat. No. A53084)

Specifications

Content And Storage Contains 50 sample cartridges, 2 positive control cartridges, and 2 negative control cartridges.
Format Kit
Sample Type Crime scene samples
For Use With (Equipment) RapidHIT™ ID
For Use With (Application) Human Identification
No. of Samples 50
Quantity 1 Kit (50 Cartridges)

For Research Use Only.