Learn More
Applied Biosystems™ RapidINTEL™ Negative Control Cartridge
The Applied Biosystems RapidINTEL Negative Control Cartridge is for use with the Applied Biosystems RapidHIT ID System to verify instrument and/or reagent performance.
Supplier:  Applied Biosystems™ P044400
Catalog No. P044400
Only null left
Explore available promotions
Description
Description
The Applied Biosystems RapidINTEL Negative Control Cartridge is for use with the Applied Biosystems RapidHIT ID System to verify instrument and/or reagent performance. Each cartridge is individually wrapped and single-use/disposable.
Required for use:
• Applied Biosystems RapidHIT ID System (Cat. No. A41810)
• Applied Biosystems RapidHIT ID Instrument Software v1.3.1 or higher
• Applied Biosystems RapidHIT ID Primary Cartridge (Cat. No. A53084)
Specifications
Specifications
Cartridge
RapidHIT™ ID System
Human Identification
1 Cartridge
Product Suggestions
Product Suggestions