missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ RapidHIT™ ID Utility Cartridge
Description
The Applied Biosystems RapidHIT ID Utility Cartridge is for use with the Applied Biosystems RapidHIT ID System during primary cartridge installation and for maintenance and service runs.
Required for use:
- Applied Biosystems RapidHIT ID System (Cat. No. A41810)
- Applied Biosystems RapidHIT ID Instrument Software v1.3.1 or higher
- Applied Biosystems RapidHIT ID Primary Cartridge (Cat. No. A53084)
Specifications
Specifications
| Format | Cartridge |
| For Use With (Equipment) | RapidHIT™ ID System |
| For Use With (Application) | Human Identification |
| Quantity | 1 cartridge |