missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ RapidHIT™ ID GlobalFiler™ Express Control (Ladder) 5-Cartridge Kit
Description
The Applied Biosystems RapidHIT ID GlobalFiler Express Control (Ladder) 5-Cartridge Kit is for use with the Applied Biosystems RapidHIT ID System. This kit contains five ACE GlobalFiler Express Control (ladder) cartridges for use with ACE GlobalFiler Express Sample cartridges.
Required for use:
- Applied Biosystems RapidHIT ID System (Cat. No. A41810)
- Applied Biosystems RapidHIT ID Instrument Software v1.3.1 or higher
- Applied Biosystems RapidHIT ID Primary Cartridge (Cat. No. A53084)
Specifications
Specifications
| Format | Kit |
| For Use With (Equipment) | RapidHIT™ ID System |
| For Use With (Application) | Human Identification |
| Quantity | 5 Cartridges |