missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ RapidHIT™ ID Booking Station ACE GlobalFiler™ Express Complete 150-Sample Kit
Description
The Applied Biosystems RapidHIT ID Booking Station ACE GlobalFiler Express Complete 150-Sample Kit is for use with the Applied Biosystems RapidHIT ID System (Cat. No. A41810) to generate lab-quality forensic DNA profiles from arrestee samples collected during the booking process. Each kit contains all the cartridges necessary to run 150 arrestee samples.
Features of the Applied Biosystems RapidHIT ID Booking Station ACE GlobalFiler Express Complete 150-Sample Kit include:
- Develop DNA profiles from arrestees in the booking station in just 90 minutes
- Upload arrestee DNA profiles to CODIS directly from the booking station
- Link arrestees to previous crimes while still in custody
Requirements:
- Applied Biosystems RapidHIT ID System (Cat. No. A41810)
- Applied Biosystems RapidHIT ID DNA Booking Station instrument software v1.0 or higher
Specifications
Specifications
| Content And Storage | • 3 x ACE GlobalFiler Express 50-Sample Kit w/ Swab (refrigerated) • Primary Cartridge Housing – GlobalFiler Express 150 (room temperature) • 2 x Utility Cartridges (room temperature) • Gel Cartridge (refrigerated) • GlobalFiler Express Control (Ladder) Cartridge (refrigerated) |
| Format | Kit |
| Sample Type | Buccal Swabs |
| For Use With (Equipment) | RapidHIT ID |
| For Use With (Application) | Booking station arrestee testing |
| No. of Samples | 150 Samples |
| Quantity | 1 Kit |