missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ RapidHIT™ ID ACE GlobalFiler™ Express Positive Control Cartridge
Description
The Applied Biosystems RapidHIT ID ACE GlobalFiler Express Positive Control Cartridge is for use with the Applied Biosystems RapidHIT ID System to verify instrument and/or reagent performance. Each cartridge is individually wrapped and single-use/disposable.
Required for use:
- Applied Biosystems RapidHIT ID System (Cat. No. A41810)
- Applied Biosystems RapidHIT ID Instrument Software v1.3.1 or higher
- Applied Biosystems RapidHIT ID Primary Cartridge (Cat. No. A53084)
Specifications
Specifications
| Format | Cartridge |
| For Use With (Equipment) | RapidHIT™ ID System |
| For Use With (Application) | Human Identification |
| Quantity | 1 cartridge |