missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ RapidHIT™ ID ACE GlobalFiler™ Express Evaluation Kit

Catalog No. 400114RH
Change view
Click to view available options
Quantity:
10 cartridges kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
400114RH 10 cartridges kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 400114RH Supplier Applied Biosystems™ Supplier No. 400114RH

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems RapidHIT ID ACE GlobalFiler Express Evaluation Kit is used with the Applied Biosystems RapidHIT ID System to generate lab-quality forensic DNA profiles primarily from reference samples collected from known individuals.

The Applied Biosystems RapidHIT ID ACE GlobalFiler Express Evaluation Kit is used with the Applied Biosystems RapidHIT ID System to generate lab-quality forensic DNA profiles primarily from reference samples collected from known individuals. These DNA profiles can be compared to DNA profiles obtained from crime scene evidence.

This kit contains 10 ACE GlobalFiler Express Sample Cartridges, each of which is an individually-wrapped, single-use, disposable cartridge that contains reagents for developing a forensic DNA profile from a single sample in 90 minutes.

To run crime scene samples on the Applied Biosystems RapidHIT ID System, see our Applied Biosystems RapidINTEL Sample Cartridge Kit (Cat No. A43941).

Intended use:
• Reference samples and high quantity crime scene samples

Required for use:
• Applied Biosystems RapidHIT ID System (Cat. No. A41810)
• Applied Biosystems RapidHIT ID Instrument Software v1.3.1 or higher
• Applied Biosystems RapidHIT ID Primary Cartridge (Cat. No. A53084)

Specifications

Format Kit
Sample Type Buccal swabs, High DNA content crime scene samples
For Use With (Equipment) RapidHIT™ ID System
For Use With (Application) Human Identification
No. of Samples 10 Samples
Quantity 10 cartridges kit