Learn More
Applied Biosystems™ RapidHIT™ ID ACE GlobalFiler™ Express 50-Sample Kit with Swab
Description
The Applied Biosystems RapidHIT ID GlobalFiler Express ACE GlobalFiler 50-Sample Kit with Swab is for use with the Applied Biosystems RapidHIT ID System to generate lab-quality forensic DNA profiles from arrestee reference samples collected during the booking process. These DNA profiles are then automatically searched against CODIS to link the arrestee to past crimes.
This kit contains 50 ACE GlobalFiler Express Sample Cartridges, each of which is an individually-wrapped, single-use, disposable cartridge that contains reagents for developing a forensic DNA profile from a single sample in 90 minutes. Included with each cartridge are swabs, tamper-proof tape, and barcodes.
Required for use:
• Applied Biosystems RapidHIT ID System (Cat. No. A41810)
• Applied Biosystems RapidHIT ID Instrument Software v1.3.1 or higher
• Applied Biosystems RapidHIT ID Primary Cartridge (Cat. No. A53084)
Specifications
Specifications
| Format | Kit |
| For Use With (Equipment) | RapidHIT™ ID System |
| For Use With (Application) | Human Identification |
| Quantity | 50 Cartridges |