Learn More
3M™ Petrifilm™ Rapid E.coli/Coliform Count Plate
Description
- E. coli and coliform results in 18 to 24 hours
- Provides accurate, efficient, consistent results for food processors and contract labs
- Plates are sample-ready, eliminating time-consuming, cumbersome step of preparing media/agar dishes
- Can be used with 3M™ Petrifilm™ Plate Reader for consistent, fast, automated reading and recording of results
- This plate has been awarded Official Methods of Analysis (OMA) #2018.13 for select food and MicroVal certificate no. 2017LR76 for a broad range of foods and select environmental surfaces by Lloyd's Register Nederland B.V. following ISO 16140-2. Food Categories tested: meat products, dairy and egg products, fish and seafood products, vegetable products, ready to eat and ready to reheat, animal feed and samples from the industrial production environment
Specifications
Specifications
| Detects | E. coli and coliform |
| Product Type | Count Plate |
| Test Type | Environmental Test |
| For Use With (Application) | Testing equipment, raw materials, food products and manufacturing environmental samples, Food processors and contract labs |
| For Use With (Equipment) | 3M™ Petrifilm™ plate reader |
| Quantity | 500 Pack |