Learn More
Promega Rapid Digestion Kit---Trypsin, 100microg DFS Item
The Rapid Trypsin and Rapid Trypsin/Lys-C Digestion Kits are designed to shorten protein digestion times to 60 minutes versus the typical 4-18 hours.
Supplier:  Promega VA1060
Catalog No. PRVA1060
Only null left
Explore available promotions
Description
Description
  • Minimize Protein Digestion Time, Maximize Results
  • Faster digestion time
  • Streamlined workflow
  • Tighter coefficients of variation
Specifications
Specifications
100 μg,100 μg
Product Suggestions
Product Suggestions