missing translation for 'onlineSavingsMsg'
Learn More

Rapeseed Oil, Spectrum™ Chemical

R2010, 8002-13-9

$1,222.31 - $1,222.31

Chemical Identifiers

CAS 8002-13-9
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18606043
View Documents
Spectrum Chemical Mfg Cor
R20104LT
4 L Amber Glass Bottle N/A
Only null left
 
18606044
View Documents
Spectrum Chemical Mfg Cor
R2010200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Rapeseed Oil
Specifications

Chemical Identifiers

8002-13-9

Specifications

8002-13-9
Amber Glass Bottle
170-180
Reagent
1
4 L
0.905-0.912
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'