missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rapeseed Oil, Spectrum™ Chemical
Description
Spectrum™ Chemical Rapeseed Oil
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 4 L |
| Saponification Value | 170-180 |
| Specific Gravity | 0.905-0.912 |
| Grade | Reagent |