missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rapamycin, 99.0%, MP Biomedicals™
$304.86 - $761.33
Chemical Identifiers
| CAS | 53123-88-9 |
|---|---|
| Molecular Formula | C51H79NO13 |
| Molecular Weight (g/mol) | 914.187 |
| InChI Key | QFJCIRLUMZQUOT-HPLJOQBZSA-N |
| Synonym | A422989, NSC226080 |
| PubChem CID | 5284616 |
| ChEBI | CHEBI:9168 |
| SMILES | CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)O)C)C)O)OC)C)C)C)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15934680
|
MP Biomedicals Inc
0215934680 |
100 μg |
Each for $304.86
|
|
|||||
|
ICN15934691
|
MP Biomedicals Inc
0215934691 |
1 mg |
Each for $761.33
|
|
|||||
Chemical Identifiers
| 53123-88-9 | |
| 914.187 | |
| A422989, NSC226080 | |
| CHEBI:9168 |
| C51H79NO13 | |
| QFJCIRLUMZQUOT-HPLJOQBZSA-N | |
| 5284616 | |
| CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)O)C)C)O)OC)C)C)C)OC |
Specifications
| Rapamycin | |
| 53123-88-9 | |
| 100 μg | |
| C51H79NO13 | |
| QFJCIRLUMZQUOT-HPLJOQBZSA-N | |
| 914.187 | |
| CHEBI:9168 | |
| Rapamycin |
| Powder | |
| White | |
| 0.99 | |
| A422989, NSC226080 | |
| CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)O)C)C)O)OC)C)C)C)OC | |
| 5284616 | |
| 914.2 |