missing translation for 'onlineSavingsMsg'
Learn More

Ranolazine Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7135701
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
R01121G 1 g
R01125G 5 g
2 options

Catalog No. R01121G

Supplier: TCI America R01121G

Only null left

Chemical Identifiers

CAS 95635-56-6
Molecular Formula C24H35Cl2N3O4
Molecular Weight (g/mol) 500.46
MDL Number MFCD03788770
InChI Key RJNSNFZXAZXOFX-UHFFFAOYNA-N
Synonym ranolazine dihydrochloride, ranolazine hcl, ranolazine hydrochloride, renolazine, n-2,6-dimethylphenyl-2-4-2-hydroxy-3-2-methoxyphenoxy propyl piperazin-1-yl acetamide dihydrochloride, ranolazine hydrochloride usan, ranolazine 2hcl, +--4-2-hydroxy-3-o-methoxyphenoxy propyl-1-piperazineaceto-2',6'-xylidide dihydrochloride
PubChem CID 71279
IUPAC Name dihydrogen N-(2,6-dimethylphenyl)-2-{4-[2-hydroxy-3-(2-methoxyphenoxy)propyl]piperazin-1-yl}acetamide dichloride
SMILES [H+].[H+].[Cl-].[Cl-].COC1=CC=CC=C1OCC(O)CN1CCN(CC(=O)NC2=C(C)C=CC=C2C)CC1

Specifications

Melting Point 227°C
Color White
Quantity 1 g
Formula Weight 500.46
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ranolazine Dihydrochloride
RTECSNumber TK7845370
TSCA No