Learn More
PeproTech RANK (soluble) Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: PeproTech 500P144BT500UG
Description
AA Sequence of recombinant protein: MQIAPPCTSE KHYEHLGRCC NKCEPGKYMS SKCTTTSDSV CLPCGPDEYL DSWNEEDKCL LHKVCDTGKA LVAVVAGNST TPRRCACTAG YHWSQDCECC RRNTECAPGL GAQHPLQLNK DTVCKPCLAG YFSDAFSSTD KCRPWTNCTF LGKRVEHHGT EKSDAVCSSS LPARK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human sRANK Receptor. Anti-Human sRANK Receptor-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human sRANK Receptor by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human sRANK Receptor (500-P144) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human sRANK Receptor. Western Blot: To detect hsRANK Receptor by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hsRANK Receptor is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P144BT-1MG will be provided as 2 x 500 μg
RANK is a member of the tumor necrosis factor receptor (TNFR) family. The full length mouse RANK cDNA encodes a type I transmembrane protein with an extracellular and a cytoplasmic domain. The extracellular domain contains two potential N-linked glycosylation sites. RANK shares significant amino acid homology with other members of the TNFR family in its extracellular four cysteine-rich repeats. Human and murine RANK share 81% amino acid identity in their extracellular domains.Chemical Identifiers
| RANK (soluble) | |
| ELISA, Western Blot | |
| Biotin | |
| Rabbit | |
| Human | |
| Q9Y6Q6 | |
| 8792 | |
| E.coli-derived, 19.3 kDa Recombinant Human sRANK Receptor. | |
| Antigen affinity chromatography | |
| TNFRSF11A |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| CD265; FEO; LOH18CR1; loss of heterozygosity, 18, chromosomal region 1; Ly109; mRANK; ODFR; OFE; OPTB7; Osteoclast differentiation factor receptor; OSTS; Paget disease of bone 2; PDB2; Rank; receptor activator of NF-kappaB; receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; RGD1563614; TNF receptor superfamily member 11a; Tnfrsf11a; TRANCER; TRANCE-R; tumor necrosis factor receptor superfamily member 11A; tumor necrosis factor receptor superfamily member 11a, NFKB activator; tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
| TNFRSF11A | |
| Primary | |
| -20°C |
Specifications
| RANK (soluble) | |
| Polyclonal | |
| Biotin | |
| TNFRSF11A | |
| CD265; FEO; LOH18CR1; loss of heterozygosity, 18, chromosomal region 1; Ly109; mRANK; ODFR; OFE; OPTB7; Osteoclast differentiation factor receptor; OSTS; Paget disease of bone 2; PDB2; Rank; receptor activator of NF-kappaB; receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; RGD1563614; TNF receptor superfamily member 11a; Tnfrsf11a; TRANCER; TRANCE-R; tumor necrosis factor receptor superfamily member 11A; tumor necrosis factor receptor superfamily member 11a, NFKB activator; tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8792 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q9Y6Q6 | |
| TNFRSF11A | |
| E.coli-derived, 19.3 kDa Recombinant Human sRANK Receptor. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |