missing translation for 'onlineSavingsMsg'
Learn More

Ranitidine Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015893
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015893 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015893 Supplier Merck Emd Millipore Supplier No. PHR1773100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Ranitidine Related Compound A is an impurity of ranitidine, an inhibitor of gastric acid secretion. Ranitidine is widely used for treating gastric and duodenal ulcers.

Specifications

CAS 91224-69-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C10H18N2 OS · 0.5 C4H4O4
Synonym 2-{{{5-[(Dimethylamino)methyl]furan-2-yl}methyl}sulfanyl}ethanamine hemifumarate salt; 2-{{{5-[(DIMETHYLAMINO)METHYL] FURAN-2-YL}METHYL}SULFANYL} ETHANEAMINEHEMIFUMARATE SALT; Ranitidine Impurity B; 2-{{{5-[(Dimethylamino)methyl]-2-furanyl}methyl}thio}ethylamine hemifumarate salt; 5-{[(2-Aminoethyl)thio]methyl}-N,N-dimethyl- 2-furanmethanamine hemifumarate salt; Ranitidine diamine hemifumarate salt
Type Certified Reference Material
Quantity 100 mg
Formula Weight 272.36
Grade Certified Reference Material