missing translation for 'onlineSavingsMsg'
Learn More

Ramipril Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012249
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012249 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012249 Supplier Merck Emd Millipore Supplier No. PHR14461G

Ramipril is an angiotensin-converting-enzyme (ACE) inhibitor, which acts on the renin angiotensin aldosterone system. It is widely used in reducing hypertension by the suppression of the effects of angiotensin II at its receptor, thereby blocking the renin-angiotensin system.

Specifications

CAS 87333-19-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C23H32N2O5
MDL Number MFCD00865775
Synonym Tritace; [2 S,3 AS,6 aS]-1-[(2 S)-2-[[(1 S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 416.51
Grade Certified Reference Material