missing translation for 'onlineSavingsMsg'
Learn More

Raloxifene Related Compound C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013435
Change view
Click to view available options
Quantity:
40 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013435 40 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013435 Supplier Merck Emd Millipore Supplier No. PHR201540MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 195454-31-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C28H27 NO5 S
Synonym 1-(2-{4-[6-Hydroxy-2-(4-hydroxyphenyl)benzothiophene-3-carbonyl]phenoxy}ethyl)piperidine 1-oxide
Type Certified Reference Material
Quantity 40 mg
Formula Weight 489.58
Grade Certified Reference Material