missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Raloxifene Hydrochloride 98.0+%, TCI America™
$103.97 - $254.11
Chemical Identifiers
| CAS | 82640-04-8 |
|---|---|
| Molecular Formula | C28H28ClNO4S |
| Molecular Weight (g/mol) | 510.045 |
| MDL Number | MFCD01938233 |
| InChI Key | BKXVVCILCIUCLG-UHFFFAOYSA-N |
| Synonym | raloxifene hydrochloride, raloxifene hcl, evista, keoxifene hydrochloride, 6-hydroxy-2-4-hydroxyphenyl benzo b thiophen-3-yl 4-2-piperidin-1-yl ethoxy phenyl methanone hydrochloride, keoxifene, unii-4f86w47br6, evista raloxifene hydrochloride, 6-hydroxy-2-p-hydroxyphenyl benzo b thien-3-yl-p-2-piperidinoethoxy phenyl ketone, hydrochloride |
| PubChem CID | 54900 |
| ChEBI | CHEBI:50740 |
| IUPAC Name | [6-hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[4-(2-piperidin-1-ylethoxy)phenyl]methanone;hydrochloride |
| SMILES | C1CCN(CC1)CCOC2=CC=C(C=C2)C(=O)C3=C(SC4=C3C=CC(=C4)O)C5=CC=C(C=C5)O.Cl |
Chemical Identifiers
| 82640-04-8 | |
| 510.045 | |
| BKXVVCILCIUCLG-UHFFFAOYSA-N | |
| 54900 | |
| [6-hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[4-(2-piperidin-1-ylethoxy)phenyl]methanone;hydrochloride |
| C28H28ClNO4S | |
| MFCD01938233 | |
| raloxifene hydrochloride, raloxifene hcl, evista, keoxifene hydrochloride, 6-hydroxy-2-4-hydroxyphenyl benzo b thiophen-3-yl 4-2-piperidin-1-yl ethoxy phenyl methanone hydrochloride, keoxifene, unii-4f86w47br6, evista raloxifene hydrochloride, 6-hydroxy-2-p-hydroxyphenyl benzo b thien-3-yl-p-2-piperidinoethoxy phenyl ketone, hydrochloride | |
| CHEBI:50740 | |
| C1CCN(CC1)CCOC2=CC=C(C=C2)C(=O)C3=C(SC4=C3C=CC(=C4)O)C5=CC=C(C=C5)O.Cl |
Specifications
| 82640-04-8 | |
| C28H28ClNO4S | |
| 1 g | |
| BKXVVCILCIUCLG-UHFFFAOYSA-N | |
| [6-hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[4-(2-piperidin-1-ylethoxy)phenyl]methanone;hydrochloride | |
| 54900 | |
| 510.05 | |
| Crystalline Powder |
| 263°C | |
| MFCD01938233 | |
| raloxifene hydrochloride, raloxifene hcl, evista, keoxifene hydrochloride, 6-hydroxy-2-4-hydroxyphenyl benzo b thiophen-3-yl 4-2-piperidin-1-yl ethoxy phenyl methanone hydrochloride, keoxifene, unii-4f86w47br6, evista raloxifene hydrochloride, 6-hydroxy-2-p-hydroxyphenyl benzo b thien-3-yl-p-2-piperidinoethoxy phenyl ketone, hydrochloride | |
| C1CCN(CC1)CCOC2=CC=C(C=C2)C(=O)C3=C(SC4=C3C=CC(=C4)O)C5=CC=C(C=C5)O.Cl | |
| 510.045 | |
| CHEBI:50740 | |
| ≥98.0% (HPLC,T) | |
| Raloxifene Hydrochloride |
Safety and Handling
RTECSNumber : PC4956925
TSCA : No