missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RAF1 (cRAF) [Y340D, Y341D], GST Tag Recombinant Protein

Catalog No. PR7307A
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR7307A 100 μg
PV3805 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR7307A Supplier Invitrogen™ Supplier No. PR7307A
Only null left

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Catalytic, Mutant.

RAF1 (cRAF) is a MAP kinase kinase (MAP3K) which functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated RAF1 can phosphorylate and activate the dual specificity protein kinases MEK1 and MEK2 which in turn phosphorylates and activates the serine/threonine specific protein kinases ERK1 and ERK2. The RAF1 Y340D Y341D mutant is constitutively active. RAF1 functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated Raf-1 can phosphorylate to activate the dual specificity protein kinases MEK1 and MEK2 which in turn phosphorylate to activate the serine/threonine specific protein kinases ERK1 and ERK2. Activated ERKs are pleiotropic effectors of cell physiology and play an important role in the control of gene expression involved in the cell division cycle, apoptosis, cell differentiation and cell migration. Mutations in this gene are associated with Noonan syndrome 5 and LEOPARD syndrome 2.

Specifications

Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 50% glycerol, 2mM DTT, 0.5mM EDTA, 150mM NaCl, 0.02% Triton X-100 and no preservative; pH 7.5
Molecular Weight (g/mol) 66.4 kDa
Name Human RAF1 (cRAF) [Y340D, Y341D], GST Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Common Name c-Raf (Y340D, Y341D Mutant)
Kinase Family RAF Kinase Family
Protein Length 306-648; Y340D, Y341D
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 306-648; Y340D, Y341D
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: TKL Group
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic, Mutant
Form Liquid
Research Category Signal Transduction
Purity or Quality Grade 80% by SDS-PAGE
Show More Show Less