Learn More
MilliporeSigma™ Supelco™ radiello™ Vertical Adapter, Threaded
Catalog No. 111010976
Change view
Click to view available options
Quantity:
20/Pk.
Catalog No. Quantity
111010976 20/Pk.
1 options
Catalog No. 111010976 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMARAD122
For personal sampling (breathing zone assessment) a vertical adapter is available to position the radiello™ diffusive body vertically on the radiello™ triangular support plate.
Specifications
For Use With (Application) Standard Use
Quantity 20/Pk.
radiello is a trademark of Institi Clinici Scientifici Maugeri