Learn More
MilliporeSigma™ Supelco™ radiello™ On-Field Thermometer
Catalog No. 111006932
Change view
Click to view available options
Quantity:
3/Pk.
Catalog No. Quantity
111006932 3/Pk.
1 options
Catalog No. 111006932 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMARAD126
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Requires no batteries, and is amenable to harsh weather conditions
Uptake rates are dependent on temperature; therefore, concentration values obtained during sampling will be more accurate if precise temperatures are recorded during sampling. Note that temperature variations of up to 4-5°C can be observed from one area of an urban population to another. The radiello™ On-Field Thermometer acts as a mini (< 1 cm3) temperature measurement station that can be mounted on the triangular support plate in conjunction with the diffusive body via a pre-attached vertical adapter. The thermometer offers a precision of +/- 5 C between -20 to 80°C, and can log up to 2048 temperature data points. This allows the researcher to record one temperature reading every 15 min. for 22 days, every 30 min. for 43 days, or 60 minutes for 85 days. The thermometer requires no batteries, and is amemable to harsh weather conditions. The radiello™ On-Field Thermometer Reader (purchased separately using RAD127) connects the thermometer to your PC (via RS232 serial port) allowing the user to program the thermometer before sampling and down load temperature readings after sampling. Each Thermometer has a unique serial number for easy identification. The software package that performs statistical and graphic analysis is NOT included with the reader.
Specifications
Includes One standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use
Quantity 3/Pk.
radiello is a trademark of Institi Clinici Scientifici Maugeri