Learn More
MilliporeSigma™ Supelco™ radiello™ Cartridge Adsorbents For Sampling HCl, Matrix SS net with silica gel (100-400 μm particle size)
Catalog No. 111009071
Change view
Click to view available options
Quantity:
20/Pk.
Catalog No. Quantity
111009071 20/Pk.
1 options
Catalog No. 111009071 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMARAD169
Only null left
For Sampling HCl
Several different radiello™ cartridge adsorbents are available. Each of which are specific for sampling different classes of compounds. The dimensions for each cartridge are 60 mm L x 4.8 or 5.8 mm D. Each cartridge is designed for one time use with the exception of radiello™ thermal desorption (TD) cartridge adsorbents. Each cartridge arrives in a sealed glass or plastic tube wrapped in a transparent thermally sealed polyethylene (PE) bag. The same sealed tube can be used to store the cartridge after sampling prior to desorption/analysis. A sufficient number of adhesive barcode labels are included with every pack of cartridges for easy tracking of sampling date and time. Note: To conduct air sampling using the radiello™ system, radiello™ diffusive bodies and triangular support plates must be ordered (separately) in addition to cartridge adsorbents. radiello™ Starter Kits (RADxxxS) include all the necessary parts to complete a sampling.
Specifications
Pore Size 100 to 400 μm
Product Line radiello
Quantity 20/Pk.
radiello is a trademark of Institi Clinici Scientifici Maugeri