missing translation for 'onlineSavingsMsg'
Learn More

Azure Biosystems Radiance Plus

Catalog No. AC2103AZ
Change view
Click to view available options
Quantity:
150 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC2103AZ 150 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC2103AZ Supplier Azure Biosystems Supplier No. AC2103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Femtogram HRP substrate for chemiluminescent Western blots (150 mL kit).

  • Radiance Plus is the most sensitive HRP substrate available for chemiluminescent Western blotting.
  • With attomole sensitivity and a long-lasting signal, Radiance Plus allows you to detect bands not visualized with other substrates. High signal-to-noise and a large dynamic range make it ideal for quantifying low-intensity bands.
  • Size: 150 mL

Specifications

Product Type ECL Western Blot Detection Reagent
Detection Method Chemiluminescence
Form Liquid
For Use With (Application) Western Blot
Quantity 150 mL
Conjugate HRP
Substrate Horseradish Peroxidase
Reactivity Chemiluminescent
Sufficient For 1500 cm2