missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAD51B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101336
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101336 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101336 Supplier Invitrogen™ Supplier No. PA5101336
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAD51L1.

Five human RAD 51 homologues have been identified: XRCC2, XRCC3, RAD51B, RAD51C, and RAD51D. Each of these homologues interacts with one or more of the others, with all of the proteins involved in one complex or multiple smaller complexes. Rad51B plays a role in meiotic recombination and/or recombinational repair.

Specifications

Antigen RAD51B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rad51b
Gene Accession No. O15315, O35719
Gene Alias AI553500; DNA repair protein RAD51 homolog 2; hREC2; MGC34245; mREC2; R51H2; R51H2REC2hREC2; RAD51 homolog B; RAD51 paralog B; Rad51B; Rad51l1; RAD51-like 1; RAD51-like protein 1; REC2; RecA-like protein; recombination repair protein; RGD1560681
Gene Symbols Rad51b
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAD51B(Accession O15315), corresponding to amino acid residues L206-T256.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19363, 5890
Target Species Human, Mouse, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less