missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac-β-Tocopherol Solution, 50 mg/mL in Hexane, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule) Vitamins, Nutraceuticals and Natural Products
Specifications
Specifications
| Packaging | Ampule |
| CAS | 148-03-8 |
| Flash Point | -23°C |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 50 mg/mL in hexane |