missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RABGGTB Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555531
Description
Immunogen sequence: MGTPQKDVII KSDAPDTLLL EKHADYIASY GSKKDDYEYC MSEYLRMSGI YWGLTVMDLM GQLHRMNREE ILAFIKSCQH ECGGIS Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 92%.
RABGGTB catalyzes the transfer of a geranyl-geranyl moiety from geranyl-geranyl pyrophosphate to both cysteines in Rab proteins with an -XXCC, -XCXC and -CCXX C-terminal, such as RAB1A, RAB3A and RAB5A respectively.Chemical Identifiers
| RABGGTB | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| RABGGTB | |
| fj12b07; geranylgeranyl transferase type II subunit beta; Geranylgeranyl transferase type-2 subunit beta; GGTase-II-beta; Ggtb; rab geranylgeranyl transferase b subunit; Rab geranylgeranyl transferase componenet, subunit beta; Rab geranylgeranyl transferase, b subunit; Rab geranylgeranyltransferase beta subunit; rab geranylgeranyltransferase subunit beta; rab geranyl-geranyltransferase subunit beta; Rab geranylgeranyltransferase, beta subunit; rab GG transferase beta; rab GGTase beta; Rabggtb; Type II protein geranyl-geranyltransferase subunit beta; wu:fj12b07; zgc:56443; zgc:56443 protein | |
| RABGGTB | |
| Primary | |
| Antigen affinity chromatography |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P53611, P53612 | |
| 19352, 5876 | |
| Recombinant protein corresponding to Human RABGGTB. Recombinant protein control fragment (Product #RP-94243). | |
| Antibody |
Specifications
| RABGGTB | |
| Polyclonal | |
| Unconjugated | |
| RABGGTB | |
| fj12b07; geranylgeranyl transferase type II subunit beta; Geranylgeranyl transferase type-2 subunit beta; GGTase-II-beta; Ggtb; rab geranylgeranyl transferase b subunit; Rab geranylgeranyl transferase componenet, subunit beta; Rab geranylgeranyl transferase, b subunit; Rab geranylgeranyltransferase beta subunit; rab geranylgeranyltransferase subunit beta; rab geranyl-geranyltransferase subunit beta; Rab geranylgeranyltransferase, beta subunit; rab GG transferase beta; rab GGTase beta; Rabggtb; Type II protein geranyl-geranyltransferase subunit beta; wu:fj12b07; zgc:56443; zgc:56443 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19352, 5876 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P53611, P53612 | |
| RABGGTB | |
| Recombinant protein corresponding to Human RABGGTB. Recombinant protein control fragment (Product #RP-94243). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |