missing translation for 'onlineSavingsMsg'
Learn More

Rabeprazole Sodium Salt 98.0+%, TCI America™

Catalog No. p-7135703
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0115100MG 100 mg
R01151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0115100MG Supplier TCI America Supplier No. R0115100MG
Only null left

Chemical Identifiers

CAS 117976-90-6
Molecular Formula C18H20N3NaO3S
Molecular Weight (g/mol) 381.43
MDL Number MFCD02092688
InChI Key KRCQSTCYZUOBHN-UHFFFAOYNA-N
PubChem CID 14720269
ChEBI CHEBI:8769
IUPAC Name sodium 2-{[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methanesulfinyl}-1H-1,3-benzodiazol-1-ide
SMILES [Na+].COCCCOC1=CC=NC(CS(=O)C2=NC3=CC=CC=C3[N-]2)=C1C

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 100 mg
Formula Weight 381.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Rabeprazole Sodium Salt
TSCA No
Recommended Storage Freezer