missing translation for 'onlineSavingsMsg'
Learn More

Rabeprazole Sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012685
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012685 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012685 Supplier Merck Emd Millipore Supplier No. PHR15251G
Pharmaceutical (small molecule)

Specifications

CAS 117976-90-6
Linear Formula C18H20N3O3 S · Na
Synonym Rabeprazole sodium; 2-([4-(3-Methoxypropoxy)-3-methylpyridin-2-yl]methylsulfinyl)-1 H-benzo[d]imidazole; Pariprazole
Type Certified Reference Material
Quantity 1 g
Formula Weight 381.42
Grade Certified Reference Material