missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Sheep IgM (H+L) Secondary Antibody [Janelia Fluor 669], R&D Systems™

Catalog No. NB783470
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB783470 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB783470 Supplier Bio-Techne Supplier No. NBP268363JF669

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

IgM (H+L) Polyclonal antibody specifically detects IgM (H+L) in Sheep samples. It is validated for ELISA

Specifications

Antigen IgM (H+L)
Applications ELISA
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Sheep IgM
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Sheep
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less