missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Sheep, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP147PMI
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP147PMI 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP147PMI Supplier MilliporeSigma Supplier No. AP147P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

This Rabbit anti-Sheep IgG Antibody, HRP conjugate is validated for use in ELISA, western blotting for the detection of Sheep IgG.

Specifications

Antigen Rabbit anti-Sheep IgG
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation RECONSTITUTION: Reconstitute with 1.5mLof sterile distilled water.
Host Species Rabbit
Purification Method Affinity Purified
Quantity 1.5 mL
Primary or Secondary Secondary
Target Species Sheep
Content And Storage 2-8°C Lyophilized for up to 12 months from date of receipt −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. Protect from light
Show More Show Less

For Research Use Only