missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Sheep, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP147BMI
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP147BMI 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP147BMI Supplier MilliporeSigma Supplier No. AP147B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Rabbit anti-Sheep IgG Antibody, biotin-SP conjugate detects level of Sheep IgG and has been published and validated for use in ELISA and western blotting.

Specifications

Antigen Rabbit anti-Sheep IgG
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation Lyophilized. Buffer = 0.01M Phosphate buffer, 0.25M NaCl with 15mg/mL BSA and 0.05% sodium azide.
Host Species Rabbit
Immunogen Purified Sheep IgG.
Purification Method Affinity Purified
Quantity 1.5 mL
Primary or Secondary Secondary
Target Species Sheep
Content And Storage 2-8°C Lyophilized for up to 12 months from date of receipt −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles..
Show More Show Less

For Research Use Only