missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Rat, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP164MI
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP164MI 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP164MI Supplier MilliporeSigma Supplier No. AP164

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Rabbit anti-Rat IgG Antibody detects level of Rat IgG and has been published and validated for use in ELISA, immunoprecipitation and western blotting.

Specifications

Antigen Rabbit anti-Rat IgG Antibody
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Purification Method Affinity Purified
Quantity 2 mg
Primary or Secondary Secondary
Target Species Rat

For Research Use Only