missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabbit anti-Rat IgG (H+L) Secondary Antibody

Catalog No. PI31218
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31218 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31218 Supplier Invitrogen™ Supplier No. LSG31218
  • ONLINE EXCLUSIVE PRICE
    $558.60
    $323.25 / Each

    Save $235.35

    42.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31218 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31218 reacts with the heavy chains of rat IgG and with the light chains common to most rat immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Rat IgG (H+L)
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.4 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.6
Host Species Rabbit
Purification Method Antigen affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less