missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabbit anti-Mouse IgM Secondary Antibody

Catalog No. PI31196
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31196 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31196 Supplier Invitrogen™ Supplier No. 31196

Rabbit Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31196 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31196 reacts with the heavy chain portion of mouse IgM, but does not react against IgG or non-immunoglobulin serum proteins based on immunoelectrophoresis. However, this antibody may cross-react with IgM from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of Origin: USA.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Mouse IgM
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.4 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.6
Host Species Rabbit
Purification Method Antigen affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less