missing translation for 'onlineSavingsMsg'
Learn More

IgMμ Rabbit anti-Mouse, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. 12488MI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12488MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12488MI Supplier MilliporeSigma Supplier No. 12488

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Rabbit anti-Mouse IgMμ Antibody, mu chain is an antibody against Mouse IgMμ for use in immunoprecipitation and western blotting.

Specifications

Antigen Rabbit anti-Mouse IgMμ
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.02M potassium phosphate, 0.15M NaCl, pH 7.2, 0.01% (w/v) sodium azide
Host Species Rabbit
Quantity 500 μg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage −20°C for up to 12 months.
Form Liquid
Show More Show Less

For Research Use Only