missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Rabbit anti-Mouse IgG2b, Clone: RM108, Secondary Antibody,

Catalog No. 041646MI
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
041646MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 041646MI Supplier MilliporeSigma™ Supplier No. 041646

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Monoclonal Antibody

Specifically detects IgG2b Clone: RM108 in Mouse samples, and it is validated for ELISA

Specifications

Antigen IgG2b
Applications ELISA
Classification Monoclonal
Clone RM108
Conjugate Unconjugated
Formulation Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.
Host Species Rabbit
Immunogen Mouse IgG2b
Quantity 100 μg
Regulatory Status RUO
Research Discipline Secondary, Control Antibodies
Primary or Secondary Secondary
Test Specificity This antibody reacts to the Fc region of mouse IgG2b. No cross reactivity with mouse IgG1, IgG2a, IgG3, IgM, IgA, IgE, human IgG, rat IgG, or goat IgG. The Fc region of RM108 has been engineered to eliminate the binding of Fc receptor.
Target Species Mouse
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only