missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Mouse IgG, Whole Molecule Polyclonal Secondary Antibody, MP Biomedicals

Catalog No. ICN55436
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55436 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55436 Supplier MP Biomedicals Inc Supplier No. 0855436
Only null left

Host Species: Rabbit
Isotype: IgG
Species Reactivity: Mouse

Specifications

Antigen IgG, Whole Molecule
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Quantity 2 mL
Primary or Secondary Secondary
Form Purified
Isotype IgG