missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Mouse, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. 06371MI
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06371MI 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06371MI Supplier MilliporeSigma Supplier No. 06371

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Rabbit anti-Mouse IgG Antibody detects level of Mouse IgG and has been published and validated for use in immunoprecipitation.

Specifications

Antigen Rabbit anti-Mouse IgG
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation of 0.02M Potassium Phosphate, pH 7.2, 0.15M NaCl, with 0.01% sodium azide
Host Species Rabbit
Immunogen Highly purified whole mouse IgG (heavy and light chains)
Purification Method Protein A chromatography
Quantity 2 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage −20°C for up to 24 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only