missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Mouse, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AQ160MI
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AQ160MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AQ160MI Supplier MilliporeSigma Supplier No. AQ160

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Rabbit anti-Mouse IgG Antibody, F(ab′)2 is an antibody against Mouse IgG for use in ELISA, immunoprecipitation and western blotting.

Specifications

Antigen Rabbit anti-Mouse IgG
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin, F(ab')2 fragment. Liquid in 0.02M Phosphate Buffer, 0.25M NaCl, 0.1% sodium azide.
Host Species Rabbit
Immunogen Highly purified mouse IgG isolated from pooled normal mouse serum.
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C for up to 6 months
Show More Show Less

For Research Use Only