missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit anti-Mouse, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP160MI
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP160MI 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP160MI Supplier MilliporeSigma Supplier No. AP160

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Detect Mouse IgG using this Rabbit anti-Mouse IgG Antibody validated for use in ELISA, immunoprecipitation and western blotting.

Specifications

Antigen Rabbit anti-Mouse IgG
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 2.4mg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen Highly purified Mouse IgG isolated from pooled normal Mouse Serum.
Quantity 2 mg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C. Do not freeze.
Form Purified
Show More Show Less

For Research Use Only